Learn More
Berlin Packaging Square Carboy with 70-400 22mm Vent Pour Cap, 5 Gallon
Catalog No. NC0870649
Encompass_Preferred
5 Gallon (640oz) Natural HDPE Square Carboy with 70-400 22mm Vent Pour Cap included, UN Rating 3H1/Y1.8/150
Catalog No. NC0870649 Supplier Berlin Packaging Supplier No. 254054
5 Gallon (640oz) Natural HDPE Square Carboy with 70-400 22mm Vent Pour Cap included, UN Rating 3H1/Y1.8/150