Learn More
Uline Jerrican, 5 Gallon, Natural
Catalog No. NC0862641
Encompass
This item must be ordered in multiples of 4 per supplier requirements
Jerrican, high density polyethylene, 5 gallon capacity, wide mouth container with screw cap, 11 x 10 x 14"
Catalog No. NC0862641 Supplier Uline Supplier No. S17471NAT
This item must be ordered in multiples of 4 per supplier requirements
Jerrican, high density polyethylene, 5 gallon capacity, wide mouth container with screw cap, 11 x 10 x 14"