Learn More
AirSea USA Salvage Drum
Catalog No. 501534835
Encompass_Preferred
Salvage Drum. 5 Gallon, holds leaks and spills of chemicals and other hazards up to 3 gallons. For transport meeting US DOT regulation 173.3 ( c )
Catalog No. 50-153-4835 Supplier AirSea USA Supplier No. SD5
Salvage Drum. 5 Gallon, holds leaks and spills of chemicals and other hazards up to 3 gallons. For transport meeting US DOT regulation 173.3 ( c )