Learn More
US Plastic Corporation 5 GALLON WHITE BUCKET
SDP
Catalog No. NC0180158
Encompass
These buckets are a great way to store your products. Made of high density polyethylene these buckets can withstand temperatures of up to 180 deg.F. Buckets are slightly tapered so you can stack them and they are easy to separate.
Catalog No. NC0180158 Supplier US Plastic Corporation Supplier No. 3826
These buckets are a great way to store your products. Made of high density polyethylene these buckets can withstand temperatures of up to 180 deg.F. Buckets are slightly tapered so you can stack them and they are easy to separate.