Learn More
AVANTOR MASTERFLEX (VWR LLC) Mag Drive PP Centrifugal Pump w/Enclosed Motor; 5 GPM/11.1 ft, 115V
Catalog No. NC0455396
Encompass
Moderate-Flow Polypropylene Pumps are an ideal option for process, recirculation, or transfer applications where continuous flow and excellent chemical resistance are needed.
Catalog No. NC0455396 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7201020
Moderate-Flow Polypropylene Pumps are an ideal option for process, recirculation, or transfer applications where continuous flow and excellent chemical resistance are needed.