Learn More
Medchemexpress LLC 5'-Guanylic acid-15N5 dilithium | 85-32-5 | 1 MG
SDP
Catalog No. 5000417728
Encompass_Preferred
5'-Guanylic acid-15N5 dilithium is a 15N labeled 5'-Guanylic acid. It is involved in several metabolic disorders, such as the AICA-ribosiduria pathway, adenosine deaminase deficiency, adenine phosphoribosyltransferase deficiency (APRT), and the 2-hydroxyglutric aciduria pathway. It is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes for quantitation during drug development
Catalog No. 50-004-17728 Supplier Medchemexpress LLC Supplier No. HYN5134S21MG
5'-Guanylic acid-15N5 dilithium is a 15N labeled 5'-Guanylic acid. It is involved in several metabolic disorders, such as the AICA-ribosiduria pathway, adenosine deaminase deficiency, adenine phosphoribosyltransferase deficiency (APRT), and the 2-hydroxyglutric aciduria pathway. It is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes for quantitation during drug development