Learn More
Medchemexpress LLC 5'-Guanylic acid-15N5 dilithium | 85-32-5 | 5 MG
SDP
Catalog No. 5000420053
Encompass_Preferred
5'-Guanylic acid-15N5 dilithium is the 15N labeled version of 5'-Guanylic acid. 5'-Guanylic acid (5'-GMP) plays a role in several metabolic disorders, including the AICA-ribosiduria pathway, adenosine deaminase deficiency, adenine phosphoribosyltransferase deficiency (aprt), and the 2-hydroxyglutric aciduria pathway. It is intended for research use only and not for sale to patients.
  • Utilized as a tracer.
  • Serves as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Stable heavy isotope.
  • Involved in metabolic pathways.
  • Intended for research use only.
Catalog No. 50-004-20053 Supplier Medchemexpress LLC Supplier No. HYN5134S25MG
Add to Cart
Add to Cart
5'-Guanylic acid-15N5 dilithium is the 15N labeled version of 5'-Guanylic acid. 5'-Guanylic acid (5'-GMP) plays a role in several metabolic disorders, including the AICA-ribosiduria pathway, adenosine deaminase deficiency, adenine phosphoribosyltransferase deficiency (aprt), and the 2-hydroxyglutric aciduria pathway. It is intended for research use only and not for sale to patients.
  • Utilized as a tracer.
  • Serves as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Stable heavy isotope.
  • Involved in metabolic pathways.
  • Intended for research use only.