Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5'-guanylic acid-15N5 dilithium | 85-32-5 | MFCD00057066 | 95.0% | 380.05 g/mol | C10H12Li215N5O8P | 10 MG
SDP

Catalog No. 5000289586
Encompass_Preferred

5'-Guanylic acid-15N5 dilithium is the 15N5 isotopically labeled dilithium salt of guanosine 5'-monophosphate supplied as a small analytical package. It is intended for use as an isotopic tracer and internal standard in quantitative analytical methods, including NMR, GC-MS, and LC-MS.

  • 15N5-labeled dilithium salt form for isotopic tracing
  • Intended as tracer and internal standard for quantitative NMR, GC-MS, and LC-MS
  • Purity 95.0% suitable for analytical applications
  • Molecular weight 380.05 g/mol
  • Provided as a 10 mg solid package for laboratory use

Catalog No. 50-002-89586 Supplier Medchemexpress LLC Supplier No. HYN5134S210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5'-Guanylic acid-15N5 dilithium is the 15N5 isotopically labeled dilithium salt of guanosine 5'-monophosphate supplied as a small analytical package. It is intended for use as an isotopic tracer and internal standard in quantitative analytical methods, including NMR, GC-MS, and LC-MS.

  • 15N5-labeled dilithium salt form for isotopic tracing
  • Intended as tracer and internal standard for quantitative NMR, GC-MS, and LC-MS
  • Purity 95.0% suitable for analytical applications
  • Molecular weight 380.05 g/mol
  • Provided as a 10 mg solid package for laboratory use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.