Learn More
Medchemexpress LLC 5'-Guanylic acid | 85-32-5 | 25MG
SDP
Catalog No. 5000421858
Encompass_Preferred
5'-Guanylic acid is a purine nucleotide that participates in physiological processes such as energy metabolism, signal transduction, and gene expression regulation. It also regulates the expression of genes related to fatty acid metabolism.
  • Weak agonist for ionotropic glutamate receptors (iGluR).
  • Reduces the activity of the glutamatergic system.
  • Exhibits neuroprotective effect.
  • Can also cause neuronal cell death at high concentrations.
Catalog No. 50-004-21858 Supplier Medchemexpress LLC Supplier No. 25MGHYN5134R
5'-Guanylic acid is a purine nucleotide that participates in physiological processes such as energy metabolism, signal transduction, and gene expression regulation. It also regulates the expression of genes related to fatty acid metabolism.
  • Weak agonist for ionotropic glutamate receptors (iGluR).
  • Reduces the activity of the glutamatergic system.
  • Exhibits neuroprotective effect.
  • Can also cause neuronal cell death at high concentrations.