Learn More
Medchemexpress LLC 5'-Guanylic acid | 85-32-5 | 50 MG
SDP
Catalog No. 5000419208
Encompass_Preferred
5'-Guanylic acid (5'-GMP) is a purine nucleotide involved in various physiological processes. It plays a role in energy metabolism, signal transduction, and gene expression regulation, particularly concerning fatty acid metabolism. It acts as a weak agonist for ionotropic glutamate receptors (iGluR), reducing the activity of the glutamatergic system and offering neuroprotective effects. However, at high concentrations, it can lead to neuronal cell death.
  • Participates in energy metabolism, signal transduction, and gene expression.
  • Regulates genes related to fatty acid metabolism.
  • Weak agonist for ionotropic glutamate receptors.
  • Exhibits neuroprotective properties.
  • Can cause neuronal cell death at high concentrations.
  • Exhibits anti-anxiety effects.
Catalog No. 50-004-19208 Supplier Medchemexpress LLC Supplier No. HYN513450MG
5'-Guanylic acid (5'-GMP) is a purine nucleotide involved in various physiological processes. It plays a role in energy metabolism, signal transduction, and gene expression regulation, particularly concerning fatty acid metabolism. It acts as a weak agonist for ionotropic glutamate receptors (iGluR), reducing the activity of the glutamatergic system and offering neuroprotective effects. However, at high concentrations, it can lead to neuronal cell death.
  • Participates in energy metabolism, signal transduction, and gene expression.
  • Regulates genes related to fatty acid metabolism.
  • Weak agonist for ionotropic glutamate receptors.
  • Exhibits neuroprotective properties.
  • Can cause neuronal cell death at high concentrations.
  • Exhibits anti-anxiety effects.