Learn More
Cayman Chemical 5-Heneicosylresorcinol, Purity: ≥95%, CAS: 70110-59-7, 10 mg
SDP
Catalog No. 502988553
Encompass_Preferred
An alkylresorcinol; reduces hydrogen peroxide-induced DNA damage in HT-29 cells at 100 µmol/L; inhibits migration and invasion, reduces the number of LC3 puncta, a marker of autophagy, and decreases survival in HepG2 cells at 160 µg/ml when used in combination with 5-nonadecanoylresorcinol
Catalog No. 50-298-8553 Supplier Cayman Chemical Supplier No. 3003110 MG
An alkylresorcinol; reduces hydrogen peroxide-induced DNA damage in HT-29 cells at 100 µmol/L; inhibits migration and invasion, reduces the number of LC3 puncta, a marker of autophagy, and decreases survival in HepG2 cells at 160 µg/ml when used in combination with 5-nonadecanoylresorcinol