Learn More
Medchemexpress LLC 5-Hepten-2-one, 6-methyl- | 110-93-0 | 99.8% | 126.20 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW010435
Encompass_Preferred
Sulcatone (6-Methyl-5-hepten-2-one) is a plant-derived volatile organic compound with insecticidal, antifungal, and blood pressure-lowering activities. It also functions as an insect pheromone and an endogenous metabolite found in feces. Changes in Sulcatone levels may aid in the auxiliary diagnosis of ulcerative colitis.
  • Plant-derived volatile organic compound
  • Insecticidal activity
  • Antifungal activity
  • Blood pressure-lowering effects
  • Insect pheromone
  • Endogenous metabolite
  • Potential auxiliary diagnostic marker for ulcerative colitis
Catalog No. 50-004-51786