Learn More
AdipoGen 5-Hexadecanoylaminofluorescein
SDP
Catalog No. 502560790
Encompass_Preferred
Chemical. CAS 73024-80-3. Formula C36H43NO6. MW 585.73. Synthetic. Lipophilic and pH-sensitive green-emitting fluorescent cell staining probe contains a C16 alkyl chain that will bind to membranes with the fluorophore at the aqueous interface and the alkyl tail protruding into the lipid interior. Useful to label plasma mebranes. Can be used to monitor surface pH by two-wavelength micro-spectrofluorometry. Used for visualization of proteins in 1-D and 2-D SDS-PAGE gels. Spectral data lambdaex=497nm, lambdaem=520nm.
Catalog No. 50-256-0790 Supplier AdipoGen Supplier No. CDXH0028M250
Chemical. CAS 73024-80-3. Formula C36H43NO6. MW 585.73. Synthetic. Lipophilic and pH-sensitive green-emitting fluorescent cell staining probe contains a C16 alkyl chain that will bind to membranes with the fluorophore at the aqueous interface and the alkyl tail protruding into the lipid interior. Useful to label plasma mebranes. Can be used to monitor surface pH by two-wavelength micro-spectrofluorometry. Used for visualization of proteins in 1-D and 2-D SDS-PAGE gels. Spectral data lambdaex=497nm, lambdaem=520nm.