Learn More
Medchemexpress LLC 5-(hexahydropyrrolo[1,2-a]pyrazine-2-carbonyl)-4-chloro-2-fluoro-N-(2-fluorophenyl)benzenesulfonamide | 1235560-28-7 | MFCD29924737 | 99.3% | 457.48 g/mol | C20H20ClF2N3O3S | 10 MG
SDP
Catalog No. 5000294773
Encompass_Preferred
ABT-639 is a peripherally acting, selective T-type Ca2+ (CaV3.2) calcium channel blocker supplied as a research reagent. It has been investigated for analgesic and antihyperalgesic activity in preclinical and clinical studies and is provided as a high-purity solid for laboratory use.
  • Peripherally acting T-type Ca2+ channel blocker.
  • Selective for CaV3.2 T-type channels.
  • High purity suitable for research applications.
  • Supplied as a solid, convenient for formulation and dosing.
  • Intended for in vitro and in vivo research use; not for human use.
Catalog No. 50-002-94773 Supplier Medchemexpress LLC Supplier No. HY1972110MG
ABT-639 is a peripherally acting, selective T-type Ca2+ (CaV3.2) calcium channel blocker supplied as a research reagent. It has been investigated for analgesic and antihyperalgesic activity in preclinical and clinical studies and is provided as a high-purity solid for laboratory use.
  • Peripherally acting T-type Ca2+ channel blocker.
  • Selective for CaV3.2 T-type channels.
  • High purity suitable for research applications.
  • Supplied as a solid, convenient for formulation and dosing.
  • Intended for in vitro and in vivo research use; not for human use.