Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hexenoic acid, 98%

Catalog No. AC449520050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC449520050 5 g
AC449520010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC449520050 Supplier Thermo Scientific Chemicals Supplier No. 449520050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 1577-22-6 | C6H10O2 | 114.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1577-22-6
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
InChI Key XUDOZULIAWNMIU-UHFFFAOYSA-N
Synonym 5-hexenoic acid, delta-hexenoic acid, acmc-1bnq8
PubChem CID 15308
IUPAC Name hex-5-enoic acid
SMILES C=CCCCC(=O)O

Specifications

Density 0.9610g/mL
Boiling Point 203.0°C
Flash Point 104°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH2=CH(CH2)3CO2H
Quantity 5 g
Specific Gravity 0.961
Solubility Information Solubility in water: insoluble.
Formula Weight 114.14
Percent Purity 98%
Chemical Name or Material 5-Hexenoic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.