Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hexyn-3-ol 90.0+%, TCI America™
SDP

Catalog No. H04625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04625ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H04625ML Supplier TCI America Supplier No. H04625ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19780-84-8
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00041593
InChI Key AJYGRAORQSCNED-UHFFFAOYSA-N
Synonym 5-hexyn-3-ol, 1-hexyn-4-ol, 4-hydroxy-1-hexyne, 4-hydroxy-1hexyne, acmc-1buup, ajygraorqscned-uhfffaoysa
PubChem CID 89223
IUPAC Name hex-5-yn-3-ol
SMILES CCC(CC#C)O

Specifications

Color Yellow
Boiling Point 139°C
Quantity 5 mL
UN Number 1987
Formula Weight 98.15
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Hexyn-3-ol
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.