Learn More
Cayman Chemical 5-HexynoIc AcId 10g
SDP
Catalog No. 502913697
Encompass_Preferred
A protein labeling compound; has been used to label proteins at acylation sites in a fatty acid synthase-dependent manner; can be further conjugated to fluorescent probes or biotin via click chemistry for protein identification
Catalog No. 50-291-3697 Supplier Cayman Chemical Supplier No. 3550710 G
A protein labeling compound; has been used to label proteins at acylation sites in a fatty acid synthase-dependent manner; can be further conjugated to fluorescent probes or biotin via click chemistry for protein identification