Learn More
Cayman Chemical 5-Hexynoic Acid, Purity: ≥95%, CAS: 53293-00-8, 5 g
SDP
Catalog No. 502910165
Encompass_Preferred
A protein labeling compound; has been used to label proteins at acylation sites in a fatty acid synthase-dependent manner; can be further conjugated to fluorescent probes or biotin via click chemistry for protein identification
Catalog No. 50-291-0165 Supplier Cayman Chemical Supplier No. 355075 G
A protein labeling compound; has been used to label proteins at acylation sites in a fatty acid synthase-dependent manner; can be further conjugated to fluorescent probes or biotin via click chemistry for protein identification