Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Hexynoic acid

Catalog No. 5440005G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-400-05G 5 g
54-400-025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 54-400-05G Supplier Sigma Aldrich Supplier No. 5440005G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 5-hexynoic acid is an alkynoic acid. it undergoes gold-catalyzed cyclization to form the corresponding methylene lactones.5-hexynoic acid may be used in the preparation of:
  • 5-hexynoic acid methyl ester
  • 5,6-dicarba-closo-dodecaboranyl hexynoic acid
  • 5,8,11-dodecatriynoic acid
  • 5-isocyanato-1-pentyne

Specifications

CAS 53293-00-8
Density 1.016 g/mL (at 25°C (literature))
Boiling Point 224°C to 225°C (lit.)
Molecular Formula C6H8O2
Refractive Index n20/D 1.449 (literature)
Linear Formula CH///C(CH2)3 CO2H
MDL Number MFCD00066346
Quantity 5 g
Molecular Weight (g/mol) 112.13
Percent Purity 97%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.