Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hexynoic acid, 96%

Catalog No. p-7028020
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2513203 1 g
AAB2513206 5 g
AAB2513214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2513203 Supplier Thermo Scientific Chemicals Supplier No. B2513203
Only null left
Add to Cart

CAS: 53293-00-8 | C6H8O2 | 112.128 g/mol

5-Hexynoic acid is used in the preparation of hex-5-ynoic acid methyl ester by using p-toluenesulfonic acid as a reagent. It is also involved in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Hexynoic acid is used in the preparation of hex-5-ynoic acid methyl ester by using p-toluenesulfonic acid as a reagent. It is also involved in chemical synthesis.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53293-00-8
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00066346
InChI Key VPFMEXRVUOPYRG-UHFFFAOYSA-N
Synonym 5-hexynoic acid, 5-hexyn-1-oic acid, 4-ethynylbutyric acid, acmc-1an4h
PubChem CID 143036
ChEBI CHEBI:73511
IUPAC Name hex-5-ynoic acid
SMILES C#CCCCC(=O)O

Specifications

Density 0.99
Boiling Point 73°C to 74°C (0.2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4495
Quantity 1 g
UN Number UN3265
Beilstein 1743192
Solubility Information Miscible with water.
Formula Weight 112.13
Percent Purity 97%
Chemical Name or Material 5-Hexynoic acid
Show More Show Less
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.