Learn More
Medchemexpress LLC 5-hexynoic acid tert-butyl ester | 73448-14-3 | MFCD13192076 | 97.0% | 168.23 g/mol | C10H16O2 | 5 G
SDP
Catalog No. 5000281668
Encompass_Preferred
tert-Butyl hex-5-ynoate is a tert-butyl ester bearing a terminal alkyne that serves as a synthetic linker and building block in organic chemistry, particularly for assembling proteolysis targeting chimera (PROTAC) molecules. The compound is listed with CAS 73448-14-3, molecular formula C10H16O2, and molecular weight 168.23 g/mol.
Catalog No. 50-002-81668 Supplier Medchemexpress LLC Supplier No. HYW4116045G
tert-Butyl hex-5-ynoate is a tert-butyl ester bearing a terminal alkyne that serves as a synthetic linker and building block in organic chemistry, particularly for assembling proteolysis targeting chimera (PROTAC) molecules. The compound is listed with CAS 73448-14-3, molecular formula C10H16O2, and molecular weight 168.23 g/mol.