Learn More
CRYSTAL TECHNOLOGY AND INDUSTRIES Frame for 500 ml Blood Bag Canister, Holds (5) C-500 Baxter Fenwal 4R9955, Origen CS500, CyroMACS 200-074-402 Canisters Frame Dimensions 8 11/16L x 31 1/16D x 9/16H" (220 x 789 x 15mm)
Catalog No. NC2208115
Encompass_Preferred
Canister Frames are constructed with corrosion resistant stainless steel and include a handle. Many sizes are available for accommodating various Canisters/Cassettes that hold the most popular brands of Blood Bag, Bone Marrow, Stem Cell, Cord Blood and Plasma Bag Storage. Custom Rack Design Services available.
This item is not returnable. View return policy
Catalog No. NC2208115 Supplier CRYSTAL TECHNOLOGY AND INDUSTRIES Supplier No. FC5005
Canister Frames are constructed with corrosion resistant stainless steel and include a handle. Many sizes are available for accommodating various Canisters/Cassettes that hold the most popular brands of Blood Bag, Bone Marrow, Stem Cell, Cord Blood and Plasma Bag Storage. Custom Rack Design Services available.