Learn More
Enzo Life Sciences 5-Hydroxymethylcytosine DNA ELISA kit (1x96wells)
Supplier: Enzo Life Sciences ADI9002250001
Highly sensitive, rapid ELISA kit to quantify 5-hydroxymethylcytosine in DNA with results that closely correlate to mass spectrometry analysis. The 5-Hydroxymethylcytosine DNA ELISA kit is a complete, colorimetric, immunometric immunoassay kit for the quantitative determination of 5-hydroxymethylcytosine in DNA with results in just 3 hours. Alternative name: 5-hmC. Sensitivity: <0.02% 5-hydroxymethylcytosine DNA per 100ng input DNA (range 0.03-0.55% (100 ng/µl)). Use/Stability: Antibody and control set should be stored at -20°C, while the DNA HRP antibody should be stored at -80°C. Assay Time: ~3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Species independent . Content: Microtiter Plate, Coating Buffer, ELISA Buffer, Primary Antibody, Antibody-conjugate, HRP Developer, Controls.