Learn More
Zymo Research Corporation 5-hmC Glucosyltransferase, 200 U
SDP
Catalog No. 502037198
Encompass_Preferred
Glucosylation of 5-hydroxymethylcytosine by 5hmC Glucosyltransferase can be used for sequence specific (see Cat. Nos. D5410 & D5411), locus specific, as well as global quantification of 5- hydroxymethylcytosine.
Catalog No. 50-203-7198 Supplier Zymo Research Corporation Supplier No. E2027
Glucosylation of 5-hydroxymethylcytosine by 5hmC Glucosyltransferase can be used for sequence specific (see Cat. Nos. D5410 & D5411), locus specific, as well as global quantification of 5- hydroxymethylcytosine.