Learn More
Epigentek Inc 5-Hydroxymethylcytosine (5-hmC) Monoclonal Antibody [HMC/4D9] (100 ug)
SDP
Supplier:  Epigentek Inc A1018100
Encompass_Preferred
Unconjugated Polyclonal Antibody Raised in Rabbit. Immunogen: Recombinant Protein of Human TET1. Tested Applications: WB, IHC, IF. Reactivity: Mouse, Rat, Human. Alternative Names: bA119F7.1 antibody, CXXC 6 antibody, CXXC finger 6 antibody, CXXC type zinc finger protein 6 antibody, KIAA1676 antibody, LCX antibody, Leukemia associated protein with a CXXC domain antibody, Methylcytosine dioxygenase TET1 antibody, Ten eleven translocation 1 antibody, Ten eleven translocation 1 gene protein homolog antibody, Tet 1 antibody, Tet methylcytosine dioxygenase 1 antibody, Tet oncogene 1 antibody, TET1 antibody, TET1_HUMAN antdy.
Catalog No. 50-109-4959