Learn More
Apexbio Technology LLC 5-hme-CTP 5 x 100 ul (100 mM)
SDP
Catalog No. 503107374
Encompass_Preferred
5-hme-CTP (5-hydroxymethylcytidine triphosphate) is a modified cytidine triphosphate bearing a hydroxymethyl group at the 5-position carbon This modification can enhance RNA stability and functionality During in vitro mRNA synthesis 5-hme-CTP can be incorporated into RNA molecules producing RNA containing 5-hydroxymethylcytidine Hydroxymethyl-modified RNA plays an essential role in studying RNA structure function and RNA-protein interactions Additionally 5-hme-CTP significantly improves RNA stability by reducing its intracellular degradation Thus this modified nucleotide has broad applications in gene expression research RNA-based drug development and gene therapy
Catalog No. 50-310-7374 Supplier Apexbio Technology LLC Supplier No. B8055.5
5-hme-CTP (5-hydroxymethylcytidine triphosphate) is a modified cytidine triphosphate bearing a hydroxymethyl group at the 5-position carbon This modification can enhance RNA stability and functionality During in vitro mRNA synthesis 5-hme-CTP can be incorporated into RNA molecules producing RNA containing 5-hydroxymethylcytidine Hydroxymethyl-modified RNA plays an essential role in studying RNA structure function and RNA-protein interactions Additionally 5-hme-CTP significantly improves RNA stability by reducing its intracellular degradation Thus this modified nucleotide has broad applications in gene expression research RNA-based drug development and gene therapy