Learn More
Apexbio Technology LLC 5-hme-UTP 100ul (100 mM)
SDP
Catalog No. 503107387
Encompass_Preferred
5-hme-UTP (5-hydroxymethyluridine triphosphate) is a modified uridine triphosphate with a hydroxymethyl group attached at the 5-carbon position This modification can enhance RNA stability and functionality During in vitro mRNA synthesis 5-hme-UTP can be incorporated into RNA molecules to generate RNA containing 5-hydroxymethyluridine RNAs modified with hydroxymethyl groups play an important role in studies of RNA structure function and interactions with proteins Additionally 5-hme-UTP significantly increases RNA stability and reduces degradation within cells Thus this modification has broad applications in gene expression research RNA therapeutic development and gene therapy
Catalog No. 50-310-7387 Supplier Apexbio Technology LLC Supplier No. B80540.1
5-hme-UTP (5-hydroxymethyluridine triphosphate) is a modified uridine triphosphate with a hydroxymethyl group attached at the 5-carbon position This modification can enhance RNA stability and functionality During in vitro mRNA synthesis 5-hme-UTP can be incorporated into RNA molecules to generate RNA containing 5-hydroxymethyluridine RNAs modified with hydroxymethyl groups play an important role in studies of RNA structure function and interactions with proteins Additionally 5-hme-UTP significantly increases RNA stability and reduces degradation within cells Thus this modification has broad applications in gene expression research RNA therapeutic development and gene therapy