Learn More
Enzo Life Sciences 5-Hydroxymethylcytosine polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADI9059040200
Encompass_Preferred
Host: Rabbit. Isotype: IgG1. Immunogen: 5-Hydroxymethylcytosine. Species reactivity: Species independent. Specificity: Recognizes 5-hydroxymethylcytosine in single-stranded DNA from any mammalian, plant, insect, or microbial sources, as well as artificial templates used for standardization. Applications: ELISA, IP Immunoblot. Purity: Purified. Formulation: Liquid. In PBS, pH 7.5, containing 0.05% sodium azide.
Catalog No. 50-200-7079