Learn More
AdipoGen 5-Hydroxymethylfluorescein diacetate (5 mg)
SDP
Supplier:  AdipoGen CDXH0145M005
Encompass_Preferred
Chemical. CAS: 143106-56-3. Formula: C25H19O8. MW: 447.108. Building block and intermediate for synthesis. Used for the synthesis of fluorescent dyes, such as 5-aminomethyl fluorescein.
Catalog No. 50-205-4491