Learn More
TARGETMOL CHEMICALS INC 5-HT1A MODULATOR 1 5MG
SDP
Catalog No. 502581755
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. 5-HT1A modulator 1 displays very high affinities for the 5-HT1A (alpha)1-adrenergic receptor and D2 receptor (IC50s = 2 nM 10 nM and 40 nM). purity: 99%
Catalog No. 50-258-1755 Supplier TARGETMOL CHEMICALS INC Supplier No. T101685MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. 5-HT1A modulator 1 displays very high affinities for the 5-HT1A (alpha)1-adrenergic receptor and D2 receptor (IC50s = 2 nM 10 nM and 40 nM). purity: 99%