Learn More
Eurofins Discoverx ChemiScreen 5-HT4B Serotonin Receptor Membrane Preparation, 1 vial. (HTS110M)
Catalog No. 502005898
Encompass
The ChemiScreen 5-HT4B Serotonin Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of HTR4B surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.
Catalog No. 50-200-5898 Supplier Eurofins Discoverx Supplier No. HTS110M
The ChemiScreen 5-HT4B Serotonin Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of HTR4B surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1 mL of 1 mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression. For detailed product information, visit www.discoverx.com/membranepreps.