Learn More
Apexbio Technology LLC 5-HYDROXY-1-TETRALONE-100MG
SDP
Catalog No. 5000570592
Encompass_Preferred
5-Hydroxy-1-tetralone (CAS No 28315-93-7) is a compound often used in biomedical research particularly in the study of neurodegenerative diseases and cancer Originating from organic synthesis its mechanism of action involves interacting with specific cellular targets and pathways that affect oxidative stress and apoptosis In vitro it displays activity typically in the nanomolar to low micromolar range which is significant for modulating enzymatic functions and receptor dynamics Applications include studies on neuronal cell lines and various cancer models where it helps in understanding disease mechanisms and potential therapeutic interventions It is also used in drug screening assays to assess efficacy and safety profiles Experimental concentrations typically depend on the specific research design allowing for flexibility in diverse scientific investigations
Catalog No. 50-005-70592 Supplier Apexbio Technology LLC Supplier No. C6662100
5-Hydroxy-1-tetralone (CAS No 28315-93-7) is a compound often used in biomedical research particularly in the study of neurodegenerative diseases and cancer Originating from organic synthesis its mechanism of action involves interacting with specific cellular targets and pathways that affect oxidative stress and apoptosis In vitro it displays activity typically in the nanomolar to low micromolar range which is significant for modulating enzymatic functions and receptor dynamics Applications include studies on neuronal cell lines and various cancer models where it helps in understanding disease mechanisms and potential therapeutic interventions It is also used in drug screening assays to assess efficacy and safety profiles Experimental concentrations typically depend on the specific research design allowing for flexibility in diverse scientific investigations