Learn More
Selleck Chemical LLC 5-Hydroxy-1-tetralone S3316-25mg
SDP
Catalog No. 503109812
Encompass_Preferred
5-hydroxy-1-tetralone is a fluorescent labeling reagent for the determination of glycosphingolipid from small amounts of biological samples
Catalog No. 50-310-9812 Supplier Selleck Chemical LLC Supplier No. S331625MG
5-hydroxy-1-tetralone is a fluorescent labeling reagent for the determination of glycosphingolipid from small amounts of biological samples