Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-hydroxy-2-methyl-7-(thiiran-2-ylmethoxy)-4H-chromen-4-one | 2133499-85-9 | 99.7% | 264.30 g·mol⁻1 | C13H12O4S | 50MG
SDP

Catalog No. 5000236619
Encompass_Preferred

HSP27 inhibitor J2 is a small-molecule research reagent that inhibits heat shock protein 27 (HSP27), inducing abnormal dimer formation and disrupting polymerization. It is intended for cellular and biochemical studies probing HSP27-related signaling and cancer biology. Handle as a research chemical and follow appropriate laboratory safety procedures.

  • High purity suitable for research applications (>99.7%).
  • CAS 2133499-85-9 for unambiguous identification.
  • Molecular formula C13H12O4S; molecular weight 264.30 g·mol⁻1.
  • Available in multiple pack sizes, including a 50 mg pack.
  • Suitable for biochemical and cellular assays targeting HSP27.
  • Store and handle according to standard laboratory safety practices.

Catalog No. 50-002-36619 Supplier Medchemexpress LLC Supplier No. HY12465350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

HSP27 inhibitor J2 is a small-molecule research reagent that inhibits heat shock protein 27 (HSP27), inducing abnormal dimer formation and disrupting polymerization. It is intended for cellular and biochemical studies probing HSP27-related signaling and cancer biology. Handle as a research chemical and follow appropriate laboratory safety procedures.

  • High purity suitable for research applications (>99.7%).
  • CAS 2133499-85-9 for unambiguous identification.
  • Molecular formula C13H12O4S; molecular weight 264.30 g·mol⁻1.
  • Available in multiple pack sizes, including a 50 mg pack.
  • Suitable for biochemical and cellular assays targeting HSP27.
  • Store and handle according to standard laboratory safety practices.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.