Learn More
Medchemexpress LLC 5-hydroxy-2-methyl-7-(thiiran-2-ylmethoxy)-4H-chromen-4-one | 2133499-85-9 | MFCD32201069 | 99.7% | 264.30 | C13H12O4S | 5MG
SDP
Catalog No. 5000237927
Encompass_Preferred
HSP27 inhibitor J2 (J2) is a small-molecule inhibitor of heat shock protein 27 (HSP27) that induces abnormal HSP27 dimer formation and inhibits formation of HSP27 giant polymers, disrupting HSP27 chaperone function and reducing cellular protection. In cellular studies J2 enhances the antiproliferative activity of 17-AAG and sensitizes cells to cisplatin, supporting its use in mechanistic and combination oncology research.
  • Inhibits HSP27 chaperone activity by promoting abnormal dimer formation.
  • Enhances antiproliferative effects of HSP90 inhibition in cell assays.
  • Sensitizes cells to platinum-based chemotherapy in vitro.
  • High purity (99.7%) suitable for biochemical and cell-based studies.
  • Supplied as milligram-scale solid packs with good DMSO solubility for assay preparation.
Catalog No. 50-002-37927 Supplier Medchemexpress LLC Supplier No. HY1246535MG
Add to Cart
Add to Cart
HSP27 inhibitor J2 (J2) is a small-molecule inhibitor of heat shock protein 27 (HSP27) that induces abnormal HSP27 dimer formation and inhibits formation of HSP27 giant polymers, disrupting HSP27 chaperone function and reducing cellular protection. In cellular studies J2 enhances the antiproliferative activity of 17-AAG and sensitizes cells to cisplatin, supporting its use in mechanistic and combination oncology research.
  • Inhibits HSP27 chaperone activity by promoting abnormal dimer formation.
  • Enhances antiproliferative effects of HSP90 inhibition in cell assays.
  • Sensitizes cells to platinum-based chemotherapy in vitro.
  • High purity (99.7%) suitable for biochemical and cell-based studies.
  • Supplied as milligram-scale solid packs with good DMSO solubility for assay preparation.