Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-hydroxy-2-methyl-7-(thiiran-2-ylmethoxy)-4H-chromen-4-one | 2133499-85-9 | MFCD32201069 | 99.7% | 264.30 g/mol | C13H12O4S | 10MG
SDP

Catalog No. 5000239059
Encompass_Preferred

HSP27 inhibitor J2 is a small-molecule research reagent that targets heat shock protein 27 (HSP27), inducing abnormal HSP27 dimer formation and disrupting polymer assembly. It is provided as a solid and as a 10 mM solution in DMSO for biochemical and cellular studies; not for clinical use.

  • Selective inhibitor of HSP27 for in vitro research.
  • High purity: 99.7%.
  • Molecular weight: 264.30 g/mol.
  • Chemical formula: C13H12O4S.
  • Available as solid and 10 mM solution in DMSO.
  • Physical form: off-white to light yellow solid.
  • Recommended storage: powder at -20°C or 4°C; in solvent at -80°C or -20°C.

Catalog No. 50-002-39059 Supplier Medchemexpress LLC Supplier No. HY12465310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HSP27 inhibitor J2 is a small-molecule research reagent that targets heat shock protein 27 (HSP27), inducing abnormal HSP27 dimer formation and disrupting polymer assembly. It is provided as a solid and as a 10 mM solution in DMSO for biochemical and cellular studies; not for clinical use.

  • Selective inhibitor of HSP27 for in vitro research.
  • High purity: 99.7%.
  • Molecular weight: 264.30 g/mol.
  • Chemical formula: C13H12O4S.
  • Available as solid and 10 mM solution in DMSO.
  • Physical form: off-white to light yellow solid.
  • Recommended storage: powder at -20°C or 4°C; in solvent at -80°C or -20°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.