Learn More
Medchemexpress LLC 5-hydroxy-3-[(2-hydroxyphenyl)methyl]-7-(2-methylnonan-2-yl)chromen-2-one | 1399049-81-0 | 98.0% | 408.53 | C26H32O4 | 10MG
SDP
Catalog No. 5000238635
Encompass_Preferred
PSB-SB-487 is a coumarin-derived research compound that acts as a potent GPR55 antagonist and CB2 receptor agonist, used in preclinical studies of metabolic disorders, neurodegeneration, neuropathic pain, and cancer. It is supplied for laboratory research in multiple small-mass quantities and includes chemical identifiers and structural data for experimental reproducibility.
  • Potent GPR55 antagonist and CB2 agonist.
  • Molecular formula C26H32O4 and molecular weight 408.53.
  • High purity (98.0%) suitable for research applications.
  • Available in small-mass sizes for laboratory experiments.
  • Relevant for studies of metabolic disorders, neurodegeneration, pain, and cancer.
  • Provided with CAS number and structural identifiers for traceability.
Catalog No. 50-002-38635 Supplier Medchemexpress LLC Supplier No. HY12511110MG
PSB-SB-487 is a coumarin-derived research compound that acts as a potent GPR55 antagonist and CB2 receptor agonist, used in preclinical studies of metabolic disorders, neurodegeneration, neuropathic pain, and cancer. It is supplied for laboratory research in multiple small-mass quantities and includes chemical identifiers and structural data for experimental reproducibility.
  • Potent GPR55 antagonist and CB2 agonist.
  • Molecular formula C26H32O4 and molecular weight 408.53.
  • High purity (98.0%) suitable for research applications.
  • Available in small-mass sizes for laboratory experiments.
  • Relevant for studies of metabolic disorders, neurodegeneration, pain, and cancer.
  • Provided with CAS number and structural identifiers for traceability.