Learn More
Medchemexpress LLC 5-hydroxy-3-[(2-hydroxyphenyl)methyl]-7-(2-methylnonan-2-yl)chromen-2-one | 1399049-81-0 | 98.0% | 408.53 g/mol | C26H32O4 | 25MG
SDP
Catalog No. 5000238691
Encompass_Preferred
PSB-SB-487 is a small-molecule research compound that functions as a potent GPR55 antagonist and a CB2 receptor agonist. It is used in preclinical pharmacology studies to investigate metabolic disorders, neurodegenerative conditions, neuropathic pain, and cancer. The compound is supplied at high purity and is characterized by CAS 1399049-81-0 and molecular formula C26H32O4.
  • Potent GPR55 antagonist (IC50 0.113 μM).
  • CB2 receptor agonist (Ki 0.292 μM for human CB2).
  • High purity suitable for preclinical research.
  • Defined molecular formula and molecular weight (C26H32O4, 408.53 g/mol).
  • Available in multiple package sizes for experimental flexibility.
Catalog No. 50-002-38691 Supplier Medchemexpress LLC Supplier No. HY12511125MG
PSB-SB-487 is a small-molecule research compound that functions as a potent GPR55 antagonist and a CB2 receptor agonist. It is used in preclinical pharmacology studies to investigate metabolic disorders, neurodegenerative conditions, neuropathic pain, and cancer. The compound is supplied at high purity and is characterized by CAS 1399049-81-0 and molecular formula C26H32O4.
  • Potent GPR55 antagonist (IC50 0.113 μM).
  • CB2 receptor agonist (Ki 0.292 μM for human CB2).
  • High purity suitable for preclinical research.
  • Defined molecular formula and molecular weight (C26H32O4, 408.53 g/mol).
  • Available in multiple package sizes for experimental flexibility.