Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-hydroxy-3-[(2-hydroxyphenyl)methyl]-7-(2-methylnonan-2-yl)chromen-2-one | 1399049-81-0 | 98.0% | 408.53 g/mol | C26H32O4 | 5MG
SDP

Catalog No. 5000238692
Encompass_Preferred

PSB-SB-487 is a small-molecule research compound that functions as a potent GPR55 antagonist and a CB2 receptor agonist. It is intended for research use only and is employed in studies of metabolic disorders, neurodegeneration, pain mechanisms, and oncology. Reported potency includes a GPR55 IC50 of 0.113 μM and a human CB2 Ki of 0.292 μM.

  • Potent GPR55 antagonist and CB2 agonist with reported potency values.
  • Molecular formula C26H32O4; molecular weight 408.53 g/mol.
  • High purity: 98.0%.
  • Appearance: white to off-white solid.
  • Recommended storage: powder at -20°C for long term, 4°C for short term; in solvent at -80°C.
  • Supplied in small milligram quantities suitable for in vitro research and pharmacology.

Catalog No. 50-002-38692 Supplier Medchemexpress LLC Supplier No. HY1251115MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PSB-SB-487 is a small-molecule research compound that functions as a potent GPR55 antagonist and a CB2 receptor agonist. It is intended for research use only and is employed in studies of metabolic disorders, neurodegeneration, pain mechanisms, and oncology. Reported potency includes a GPR55 IC50 of 0.113 μM and a human CB2 Ki of 0.292 μM.

  • Potent GPR55 antagonist and CB2 agonist with reported potency values.
  • Molecular formula C26H32O4; molecular weight 408.53 g/mol.
  • High purity: 98.0%.
  • Appearance: white to off-white solid.
  • Recommended storage: powder at -20°C for long term, 4°C for short term; in solvent at -80°C.
  • Supplied in small milligram quantities suitable for in vitro research and pharmacology.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.