Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hydroxy-3,4-dimethoxybenzoic acid, 97+%

Catalog No. AAL05708MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL05708MD 250 mg
AAL0570803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL05708MD Supplier Thermo Scientific Chemicals Supplier No. L05708MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 8-1-1916 | C9H10O5 | 198.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8-1-1916
Molecular Formula C9H10O5
Molecular Weight (g/mol) 198.17
MDL Number MFCD00002508
InChI Key WFIBQVFJXGQICQ-UHFFFAOYSA-N
Synonym 3,4-dimethoxy-5-hydroxybenzoic acid, 5-hydroxy-3,4-dimethoxybenzoic acid, 5-hydroxyveratric acid, 3,4-o-dimethylgallic acid, 5-hydroxy veratric acid, chembl85234, 3,4-dimethoxy-5-hydroxy benzoic acid, acmc-20an2p, 3-hydroxy-4 5-dimethoxybenzoic acid, 3-hydroxy-4,5-dimethoxy-benzoic acid
PubChem CID 74709
IUPAC Name 3-hydroxy-4,5-dimethoxybenzoic acid
SMILES COC1=CC(=CC(=C1OC)O)C(=O)O

Specifications

Melting Point 194°C to 197°C
Quantity 250 mg
Beilstein 2115310
Formula Weight 198.17
Percent Purity ≥97%
Chemical Name or Material 5-Hydroxy-3,4-dimethoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-630-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.