Learn More
Medchemexpress LLC 5-hydroxy-3-(indol-3-yl)isoindolin-1-one (S-isomer) | 2375482-51-0 | 99.5% | 516.59 Da | C31H28N6O2 | 50MG
SDP
Catalog No. 5000239249
Encompass_Preferred
BI-2852 is a small-molecule inhibitor that binds the KRAS switch I/II pocket and inhibits active, GTP-bound KRAS with nanomolar potency. It is intended for research use and is used as a tool compound to study KRAS-driven signaling, pERK modulation, and antiproliferative effects in KRAS-mutant cell models.
  • Potent nanomolar inhibition of KRAS switch I/II pocket.
  • Directly targets active GTP-bound KRAS to disrupt effector interactions.
  • Suitable for in vitro studies of KRAS signaling and cell proliferation.
  • High purity (>99%) suitable for biochemical and cell-based assays.
  • Molecular formula C31H28N6O2 and molecular weight 516.59 Da.
  • Available with supporting documentation, including COA and SDS.
Catalog No. 50-002-39249 Supplier Medchemexpress LLC Supplier No. HY12624750MG
BI-2852 is a small-molecule inhibitor that binds the KRAS switch I/II pocket and inhibits active, GTP-bound KRAS with nanomolar potency. It is intended for research use and is used as a tool compound to study KRAS-driven signaling, pERK modulation, and antiproliferative effects in KRAS-mutant cell models.
  • Potent nanomolar inhibition of KRAS switch I/II pocket.
  • Directly targets active GTP-bound KRAS to disrupt effector interactions.
  • Suitable for in vitro studies of KRAS signaling and cell proliferation.
  • High purity (>99%) suitable for biochemical and cell-based assays.
  • Molecular formula C31H28N6O2 and molecular weight 516.59 Da.
  • Available with supporting documentation, including COA and SDS.