Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-hydroxy-4H-1,4-benzoxazin-3-one derivative (substituted amine) | 1202235-68-4 | 99.6% | 25 MG
SDP

Catalog No. 5000233483
Encompass_Preferred

BI-167107 is a high-affinity full agonist of the β2 adrenergic receptor (β2AR) with a reported dissociation constant of approximately 84 pM. Supplied as a high-purity solid for research use, it is commonly used as a pharmacology and structural biology tool because of its slow dissociation rate and strong receptor affinity.

  • High-affinity β2AR full agonist with Kd ≈ 84 pM.
  • High purity solid suitable for biochemical assays and structural studies.
  • Available in small milligram pack sizes for research use.
  • Soluble in DMSO; available as a 10 mM solution format for assay preparation.
  • Molecular weight 370.4 g/mol; formula C21H26N2O4.
  • Used as a tool compound for receptor pharmacology and crystallography due to very slow off-rate.

Catalog No. 50-002-33483 Supplier Medchemexpress LLC Supplier No. HY12125125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BI-167107 is a high-affinity full agonist of the β2 adrenergic receptor (β2AR) with a reported dissociation constant of approximately 84 pM. Supplied as a high-purity solid for research use, it is commonly used as a pharmacology and structural biology tool because of its slow dissociation rate and strong receptor affinity.

  • High-affinity β2AR full agonist with Kd ≈ 84 pM.
  • High purity solid suitable for biochemical assays and structural studies.
  • Available in small milligram pack sizes for research use.
  • Soluble in DMSO; available as a 10 mM solution format for assay preparation.
  • Molecular weight 370.4 g/mol; formula C21H26N2O4.
  • Used as a tool compound for receptor pharmacology and crystallography due to very slow off-rate.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.