Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-hydroxy-8-(1-hydroxy-2-((2-methyl-1-(o-tolyl)propan-2-yl)amino)ethyl)-4H-benzoxazin-3-one | 1202235-68-4 | 99.6% | 1 ML
SDP

Catalog No. 5000233385
Encompass_Preferred

BI-167107 is a research-grade small-molecule full agonist of the β2 adrenergic receptor used as a pharmacological tool in receptor activation, signaling, and structural biology studies. It is supplied for biochemical and biophysical experiments and is typically provided at high purity.

  • High-affinity full agonist for β2 adrenergic receptor (Kd ≈ 84 pM).
  • Suitable for receptor pharmacology, crystallography, and signaling assays.
  • Molecular formula C21H26N2O4; molecular weight 370.4 g/mol.
  • High purity (≈99.6%) and white to off-white solid appearance.
  • Stable when stored at -20°C (powder) and -80°C (solutions).

Catalog No. 50-002-33385 Supplier Medchemexpress LLC Supplier No. HY12125110MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BI-167107 is a research-grade small-molecule full agonist of the β2 adrenergic receptor used as a pharmacological tool in receptor activation, signaling, and structural biology studies. It is supplied for biochemical and biophysical experiments and is typically provided at high purity.

  • High-affinity full agonist for β2 adrenergic receptor (Kd ≈ 84 pM).
  • Suitable for receptor pharmacology, crystallography, and signaling assays.
  • Molecular formula C21H26N2O4; molecular weight 370.4 g/mol.
  • High purity (≈99.6%) and white to off-white solid appearance.
  • Stable when stored at -20°C (powder) and -80°C (solutions).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.