Learn More
Cayman Chemical 5-hydroxy Diclofenac, Purity: ≥95%, CAS: 69002-84-2, 5 mg
SDP
Catalog No. 502993703
Encompass_Preferred
An active metabolite of diclofenac; cytotoxic to HL-60 leukemia cells (IC50 = 661 µM); induces apoptosis and increases caspase-3 activity in rat hepatocytes in a concentration-dependent manner
Catalog No. 50-299-3703 Supplier Cayman Chemical Supplier No. 233695 MG
An active metabolite of diclofenac; cytotoxic to HL-60 leukemia cells (IC50 = 661 µM); induces apoptosis and increases caspase-3 activity in rat hepatocytes in a concentration-dependent manner