Learn More
Cayman Chemical 5'-hydroxy Meloxicam, Purity: ≥98%, CAS: 130262-92-9, 1 mg
SDP
Catalog No. 502985695
Encompass_Preferred
A metabolite of meloxicam; formed via metabolism of meloxicam primarily by CYP2C9 and to a lesser extent by CYP3A4
Catalog No. 50-298-5695 Supplier Cayman Chemical Supplier No. 221591 MG
A metabolite of meloxicam; formed via metabolism of meloxicam primarily by CYP2C9 and to a lesser extent by CYP3A4