Learn More
Cayman Chemical 5-hydroxy MeloxIcam 500ug
SDP
Catalog No. 502987724
Encompass_Preferred
A metabolite of meloxicam; formed via metabolism of meloxicam primarily by CYP2C9 and to a lesser extent by CYP3A4
Catalog No. 50-298-7724 Supplier Cayman Chemical Supplier No. 22159500 UG
A metabolite of meloxicam; formed via metabolism of meloxicam primarily by CYP2C9 and to a lesser extent by CYP3A4