Learn More
Cayman Chemical 5-hydroxy ThIabendazole 10mg
SDP
Catalog No. 502932646
Encompass_Preferred
A major metabolite of thiabendazole; has no effect on the growth of third-stage A. caninum larvae
Catalog No. 50-293-2646 Supplier Cayman Chemical Supplier No. 2502110 MG
A major metabolite of thiabendazole; has no effect on the growth of third-stage A. caninum larvae