Learn More
AdipoGen 5-Hydroxydiclofenac
SDP
Supplier:  AdipoGen CDXH0053M005
Encompass_Preferred
Chemical. CAS 69002-84-2. Formula C14H11Cl2NO3. MW 312.2. Synthetic. Metabolite of Diclofenac, a nonsteroidal anti-inflammatory compound that functions via inhibiton of cyclooxygenase COX. Diclofenac undergoes biotransformation not only by glucuronidation, but also by cytochrome P450–mediated oxidation to produce 4-OH-DFN and 5-OH-DFN, which undergo subsequent oxidation to form highly electrophilic benzoquinone imine intermediates. Compound can be used as analytical reference material.
Catalog No. 50-256-0774