Learn More
AdipoGen 5-Hydroxyferulic acid
SDP
Catalog No. 502561026
Encompass_Preferred
Chemical. CAS 1782-55-4. Formula C10H10O5. MW 210.18. Synthetic Hydroxycinnamic acid. Secondary plant metabolite of the phenylpropanoid pathway. It is a precursor in the biosynthesis of sinapic acid. Formed from ferulic acid by the action of the specific enzyme ferulate 5-hydroxylase F5H.
Catalog No. 50-256-1026 Supplier AdipoGen Supplier No. CDXD0915M100
Chemical. CAS 1782-55-4. Formula C10H10O5. MW 210.18. Synthetic Hydroxycinnamic acid. Secondary plant metabolite of the phenylpropanoid pathway. It is a precursor in the biosynthesis of sinapic acid. Formed from ferulic acid by the action of the specific enzyme ferulate 5-hydroxylase F5H.