Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-hydroxyferulic acid | 1782-55-4 | 99.3% | C10H10O5 | 10MG
SDP

Catalog No. 5000261677
Encompass_Preferred

5-Hydroxyferulic acid is a hydroxycinnamic acid and metabolite used as a biochemical intermediate and research reagent. It functions as a precursor in sinapic acid biosynthesis and as a substrate in catechol-O-methyltransferase (COMT) studies. The compound is provided as a solid suitable for analytical and small-scale laboratory work.

  • Purity 99.3%.
  • Molecular formula C10H10O5.
  • Molecular weight 210.18 g/mol.
  • Physical form solid; melting point 182 °C.
  • Available in mg-scale quantities for research use.
  • Useful as a biosynthetic precursor and COMT substrate.

Catalog No. 50-002-61677 Supplier Medchemexpress LLC Supplier No. HY13306810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Hydroxyferulic acid is a hydroxycinnamic acid and metabolite used as a biochemical intermediate and research reagent. It functions as a precursor in sinapic acid biosynthesis and as a substrate in catechol-O-methyltransferase (COMT) studies. The compound is provided as a solid suitable for analytical and small-scale laboratory work.

  • Purity 99.3%.
  • Molecular formula C10H10O5.
  • Molecular weight 210.18 g/mol.
  • Physical form solid; melting point 182 °C.
  • Available in mg-scale quantities for research use.
  • Useful as a biosynthetic precursor and COMT substrate.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.